Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129981.2 | NP_001123453.1 | 7 | 1049 | ||
NM_001291218.1 | NP_001278147.1 | 8 | 1148 | |||
NM_001291219.1 | NP_001278148.1 | 7 | 1049 | |||
NM_020349.3 | NP_065082.2 | 8 | 1148 | |||
GenBank mRNA | AB058599.1 | - | 8 | 810 | ||
AJ304805.1 | - | 8 | 810 | |||
AJ346356.1 | - | 2 | 84 | |||
AJ583444.1 | - | 8 | 1148 | |||
AK056990.1 | - | 8 | 1090 | |||
AK314763.1 | - | 8 | 810 | |||
BC020817.1 | - | 7 | 711 | |||
BC107759.1 | - | 8 | 1051 | 4096546 |