Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001240.3 | NP_001231.2 | 3 | 591 | ||
NM_001277842.1 | NP_001264771.1 | 3 | 591 | |||
XM_017020197.1 | XP_016875686.1 | 2 | 102 | |||
GenBank mRNA | AF045161.1 | - | 3 | 268 | ||
AF048730.1 | - | 3 | 312 | |||
AK290990.1 | - | 3 | 491 | |||
EF688064.1 | - | 3 | 268 |