Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012333.4 | NP_036465.2 | 2, 3 | 167 | ||
GenBank mRNA | AB007191.2 | - | 2, 3 | 132 | ||
AB451287.1 | - | 2, 3 | 94 | |||
AB451419.1 | - | 2, 3 | 94 | |||
AK311886.1 | - | 2, 3 | 157 | |||
BC008686.1 | - | 2, 3 | 132 | 3870100 | ||
BT007109.1 | - | 2, 3 | 94 | |||
BU601195.1 | - | 2, 3 | 169 | |||
CR456936.1 | - | 2, 3 | 94 | |||
D50692.1 | - | 2, 3 | 140 |
Gene Symbol: GJA9-MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037633.1 | 3, 4 | 2873 | |||
NR_037634.1 | 4, 5 | 2005 | ||||
NR_037635.1 | 2, 3 | 274 | ||||
GenBank mRNA | AK123051.1 | - | 4, 5 | 1999 | ||
BC039090.1 | - | 2, 3 | 894 | |||
BC051675.1 | - | 3, 4 | 2845 | |||
FY211295.1 | - | 2, 3 | 250 |