Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206615.1 | NP_001193544.1 | 7 | 796 | ||
NM_001206616.1 | NP_001193545.1 | 8 | 866 | |||
NM_012153.5 | NP_036285.2 | 8 | 865 | |||
XM_005252857.1 | XP_005252914.1 | 8 | 863 | |||
XM_005252860.1 | XP_005252917.1 | 8 | 719 | |||
XM_005252861.1 | XP_005252918.1 | 7 | 797 | |||
XM_005252862.1 | XP_005252919.1 | 7 | 666 | |||
XM_011519983.1 | XP_011518285.1 | 8 | 891 | |||
XM_011519984.1 | XP_011518286.1 | 9 | 971 | |||
XM_011519985.1 | XP_011518287.1 | 8 | 756 | |||
XM_017017534.1 | XP_016873023.1 | 7 | 794 | |||
GenBank mRNA | AF124438.1 | - | 7 | 702 | ||
AF124439.1 | - | 8 | 771 | |||
AF170583.1 | - | 8 | 775 | |||
AF203977.1 | - | 8 | 786 | |||
AF212848.1 | - | 8 | 805 | |||
AK309318.1 | - | 8 | 891 | |||
AK310867.1 | - | 8 | 866 | |||
AY882601.1 | - | 7 | 674 | |||
BC038995.2 | - | 8 | 825 | 6055934 |