Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR5K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004736.3 | NP_001004736.2 | 1 | 672 | ||
GenBank mRNA | BC136918.1 | - | 1 | 672 | 9020909 | |
BC136919.1 | - | 1 | 672 | 9020910 | ||
X64984.1 | - | 1 | 201 |