Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM162A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014367.3 | NP_055182.3 | 2 | 178 | ||
GenBank mRNA | AF107495.1 | - | 2 | 159 | ||
AF191020.1 | - | 2 | 141 | |||
AF201944.1 | - | 2 | 171 | |||
AF250321.1 | - | 2 | 158 | |||
AK026030.1 | - | 2 | 172 | |||
AK293994.1 | - | 2 | 156 | |||
BC010896.1 | - | 2 | 176 | 4283379 | ||
BC015060.1 | - | 2 | 176 | 4248082 | ||
BI460077.1 | - | 3 | 343 |