Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013451.3 | NP_038479.1 | 38 | 4408 | ||
NM_133337.2 | NP_579899.1 | 37 | 4369 | |||
XM_005269693.4 | XP_005269750.1 | 38 | 4587 | |||
XM_005269694.4 | XP_005269751.1 | 37 | 4548 | |||
XM_011539632.2 | XP_011537934.1 | 39 | 4781 | |||
XM_017016068.1 | XP_016871557.1 | 37 | 4168 | |||
XM_017016069.1 | XP_016871558.1 | 37 | 4213 | |||
XM_017016070.1 | XP_016871559.1 | 38 | 4595 | |||
GenBank mRNA | AB033033.2 | - | 38 | 4285 | ||
AF182316.1 | - | 38 | 4381 | |||
AF207990.1 | - | 37 | 4246 | |||
AK075258.1 | - | 2 | 123 | |||
AL096713.1 | - | 18 | 2425 |