Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012458.3 | NP_036590.1 | 2 | 526 | ||
GenBank mRNA | AF144700.1 | - | 2 | 189 | ||
AF152352.1 | - | 2 | 143 | |||
AK024764.1 | - | 2 | 161 | |||
BC008607.1 | - | 2 | 184 | 3453921 | ||
BI908100.1 | - | 2 | 487 | |||
DA890574.1 | - | 2 | 498 | |||
DA947460.1 | - | 2 | 533 |