Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012456.2 | NP_036588.1 | 3 | 471 | ||
GenBank mRNA | AF150089.1 | - | 3 | 453 | ||
AK289751.1 | - | 3 | 422 | |||
BC032133.2 | - | 3 | 434 | 4761702 |