Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012275.2 | NP_036407.1 | 3 | 210 | ||
NM_173170.1 | NP_775262.1 | 3 | 182 | |||
GenBank mRNA | AF186094.1 | - | 3 | 211 | ||
AF201830.1 | - | 2 | 76 | |||
AF230377.1 | - | 2 | 49 | |||
AJ242737.1 | - | 3 | 130 | |||
AJ242738.1 | - | 3 | 121 | |||
AK290249.1 | - | 3 | 120 | |||
AY359117.1 | - | 3 | 115 | |||
BC024747.1 | - | 3 | 182 | 4996939 |