Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012275.2 | NP_036407.1 | 4 | 391 | ||
NM_173170.1 | NP_775262.1 | 4 | 363 | |||
GenBank mRNA | AF186094.1 | - | 4 | 392 | ||
AF201830.1 | - | 3 | 257 | |||
AF230377.1 | - | 3 | 230 | |||
AJ242737.1 | - | 4 | 311 | |||
AJ242738.1 | - | 4 | 302 | |||
AK290249.1 | - | 4 | 301 | |||
AY359117.1 | - | 4 | 296 | |||
BC024747.1 | - | 4 | 363 | 4996939 |