Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191322.1 | NP_001178251.1 | 3 | 225 | ||
NM_001191323.1 | NP_001178252.1 | 3 | 312 | |||
NM_013372.6 | NP_037504.1 | 2 | 435 | |||
XM_005254301.2 | XP_005254358.1 | 2 | 499 | |||
XM_017022077.1 | XP_016877566.1 | 2 | 481 | |||
GenBank mRNA | AF045800.1 | - | 1 | 276 | ||
AF110137.2 | - | 2 | 338 | |||
AF154054.1 | - | 2 | 405 | |||
AK095741.1 | - | 2 | 279 | |||
AK095890.1 | - | 3 | 312 | |||
AK095972.1 | - | 3 | 225 | |||
AK312265.1 | - | 2 | 436 | |||
AY232290.1 | - | 2 | 411 | |||
BC069525.1 | - | 1 | 276 | 7262108 | ||
BC093778.1 | - | 2 | 298 | 7939623 | ||
BC101611.1 | - | 2 | 298 | 8069117 |