Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288725.1 | NP_001275654.1 | 9 | 1191 | ||
NM_001288726.1 | NP_001275655.1 | 9 | 1188 | |||
NM_019040.4 | NP_061913.3 | 9 | 1188 | |||
GenBank mRNA | AK000505.1 | - | 9 | 1167 | ||
AK304885.1 | - | 9 | 1150 | |||
AL136677.1 | - | 9 | 1145 | |||
BC012514.1 | - | 9 | 1139 | 4509903 |