Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012345.2 | NP_036477.2 | 6 | 849 | ||
GenBank mRNA | AF159548.1 | - | 6 | 807 | ||
AK314242.1 | - | 6 | 842 | |||
BC017745.1 | - | 4 | 332 | 4398678 |