Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPS6KC1 SIRNA Assays ›
Gene Symbol:
RPS6KC1
Gene Name:
ribosomal protein S6 kinase C1
Gene Aliases:
RPK118, RSKL1, S6K-delta-1, S6PKh1, humS6PKh1
Chromosome Location:
Chr.1: 213051233 - 213378753 on Build GRCh38
UniGene ID:
Hs.591416
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s25625
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RPS6KC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136138.2
NP_001129610.1
9, 10
1372
NM_001287218.1
NP_001274147.1
10, 11
1393
NM_001287219.1
NP_001274148.1
10, 11
1432
NM_001287220.1
NP_001274149.1
8, 9
1227
NM_001287221.1
NP_001274150.1
9, 10
1287
NM_012424.4
NP_036556.2
10, 11
1408
XM_005273095.1
XP_005273152.1
9, 10
1302
XM_005273096.1
XP_005273153.1
11, 12
1506
XM_006711272.1
XP_006711335.1
9, 10
1032
XM_017001024.1
XP_016856513.1
8, 9
1069
XM_017001025.1
XP_016856514.1
11, 12
1459
XM_017001026.1
XP_016856515.1
11, 12
1452
XM_017001027.1
XP_016856516.1
12, 13
1584
XM_017001028.1
XP_016856517.1
11, 12
1491
XM_017001029.1
XP_016856518.1
10, 11
1143
XM_017001030.1
XP_016856519.1
12, 13
1657
XM_017001031.1
XP_016856520.1
13, 14
1735
XM_017001032.1
XP_016856521.1
9, 10
1280
XM_017001033.1
XP_016856522.1
9, 10
1325
XM_017001034.1
XP_016856523.1
10, 11
1403
GenBank mRNA
AB070706.1
-
10, 11
1236
AF477978.1
-
10, 11
1385
AK122921.1
-
9, 10
1343
AK122989.1
-
10, 11
1395
AK295053.1
-
7, 8
886
AK299298.1
-
10, 11
1418
BC104769.1
-
10, 11
1322
8143772
BC143473.1
-
8, 9
1141
BX647303.1
-
8, 9
1050
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00179396_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction
protein serine/threonine kinase activity
protein binding
ATP binding
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline