Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPLAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017570.4 | NP_060040.1 | 10 | 1278 | ||
XM_011516960.1 | XP_011515262.1 | 10 | 1554 | |||
GenBank mRNA | AB122018.1 | - | 9 | 1174 | ||
AL096750.1 | - | 4 | 633 | |||
BC036944.1 | - | 5 | 465 | |||
BC142672.1 | - | 10 | 1278 | 40148181 | ||
BC150206.1 | - | 10 | 1256 | 40148883 |