Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPLAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017570.4 | NP_060040.1 | 18 | 2602 | ||
XM_011516960.1 | XP_011515262.1 | 18 | 2878 | |||
GenBank mRNA | AB122018.1 | - | 17 | 2498 | ||
AF217994.2 | - | 150 | ||||
AL096750.1 | - | 10 | 1587 | |||
BC036944.1 | - | 13 | 1789 | |||
BC041576.1 | - | 3 | 413 | |||
BC142672.1 | - | 18 | 2602 | 40148181 | ||
BC150206.1 | - | 18 | 2580 | 40148883 |