Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK223393.1 | - | 4 | 656 | ||
AK301154.1 | - | 4 | 679 | |||
AK303367.1 | - | 4 | 480 | |||
AK310676.1 | - | 4 | 518 | |||
AK313614.1 | - | 4 | 685 | |||
BC033583.1 | - | 4 | 640 | 5227138 | ||
CR996443.1 | - | 4 | 740 | |||
M14362.1 | - | 4 | 636 | |||
M16336.1 | - | 4 | 650 | |||
M16445.1 | - | 4 | 633 |