Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290033.1 | NP_001276962.1 | 4 | 283 | ||
NM_012133.5 | NP_036265.3 | 4 | 283 | |||
GenBank mRNA | AB047847.1 | - | 4 | 193 | ||
AF157833.1 | - | 4 | 253 | |||
AF207598.1 | - | 4 | 249 | |||
AK022934.1 | - | 4 | 180 | |||
AK291409.1 | - | 4 | 273 | |||
BC017443.1 | - | 4 | 176 | |||
BC110796.1 | - | 4 | 230 | 5197024 | ||
BG470623.1 | - | 4 | 176 | |||
BG704011.1 | - | 4 | 249 | |||
BX353590.2 | - | 4 | 283 |