Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291488.1 | NP_001278417.1 | 5 | 766 | ||
NM_001291489.1 | NP_001278418.1 | 4 | 836 | |||
NM_001291490.1 | NP_001278419.1 | 5 | 900 | |||
NM_001291491.1 | NP_001278420.1 | 3 | 618 | |||
NM_152354.5 | NP_689567.4 | 4 | 745 | |||
GenBank mRNA | AK055309.1 | - | 1 | 354 | ||
AL833447.1 | - | 4 | 723 | |||
AY166790.1 | - | 2 | 535 | |||
BC069384.1 | - | 1 | 264 | 7262358 | ||
BC074824.2 | - | 1 | 227 | |||
BC074875.2 | - | 1 | 227 | 30915235 | ||
BC117300.1 | - | 3 | 709 | 40125851 | ||
BC117302.1 | - | 3 | 709 | 40125853 | ||
BC143984.1 | - | 4 | 730 |