Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP8L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291478.1 | NP_001278407.1 | 2 | 249 | ||
NM_014371.3 | NP_055186.3 | 2 | 249 | |||
NR_111971.1 | 2 | 249 | ||||
XM_005259854.3 | XP_005259911.2 | 2 | 156 | |||
XM_011527916.2 | XP_011526218.1 | 2 | 156 | |||
GenBank mRNA | AB015332.1 | - | 2 | 96 | ||
AB025905.1 | - | 2 | 96 | |||
AB451326.1 | - | 1 | 51 | |||
AB451469.1 | - | 1 | 51 | |||
AF199414.1 | - | 1 | 51 | |||
AJ243467.1 | - | 2 | 97 | |||
AK001594.1 | - | 2 | 116 | |||
AK094736.1 | - | 2 | 116 | |||
AK225046.1 | - | 2 | 116 | |||
AK225291.1 | - | 2 | 116 | |||
AK295956.1 | - | 2 | 154 | |||
BC000713.2 | - | 2 | 101 | 3504388 | ||
BI458860.1 | - | 2 | 154 | |||
CD244113.1 | - | 2 | 100 | |||
DA315789.1 | - | 2 | 233 | |||
DB099586.1 | - | 2 | 249 |