Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014379.3 | NP_055194.1 | 4 | 2112 | ||
GenBank mRNA | AF167082.2 | - | 3 | 1420 | ||
AK297393.1 | - | 4 | 1186 | |||
AK308123.1 | - | 4 | 2105 | |||
AK313469.1 | - | 3 | 1412 | |||
BC028739.2 | - | 3 | 1386 | 5261372 |