Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321389.1 | NP_001308318.1 | 3 | 512 | ||
NM_001321390.1 | NP_001308319.1 | 3 | 157 | |||
NM_001321391.1 | NP_001308320.1 | 3 | 191 | |||
NM_015680.5 | NP_056495.3 | 2 | 249 | |||
NR_135628.1 | 3 | 191 | ||||
NR_135629.1 | 2 | 249 | ||||
GenBank mRNA | AF151815.1 | - | 3 | 416 | ||
AK075274.1 | - | 2 | 205 | |||
AK311656.1 | - | 2 | 186 | |||
AK314975.1 | - | 2 | 192 | |||
AY007160.1 | - | 2 | 222 | |||
BC001393.2 | - | 2 | 172 | 3051397 | ||
BC012821.2 | - | 2 | 221 | 2958621 | ||
BI912613.1 | - | 3 | 158 | |||
CR457401.1 | - | 2 | 84 | |||
CX166359.1 | - | 3 | 508 | |||
DB477110.1 | - | 2 | 236 | |||
HY106884.1 | - | 2 | 249 |