Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282674.1 | NP_001269603.1 | 2 | 735 | ||
NM_014380.2 | NP_055195.1 | 1 | 726 | |||
NM_206915.2 | NP_996798.1 | 3 | 814 | |||
NM_206917.2 | NP_996800.1 | 3 | 767 | |||
GenBank mRNA | AF187064.1 | - | 1 | 726 | ||
AI929106.1 | - | 2 | 510 | |||
AK057859.1 | - | 3 | 587 | |||
AY833562.1 | - | 3 | 611 | |||
BC003190.1 | - | 3 | 610 | 3357965 | ||
BC051713.1 | - | 1 | 1406 | |||
BQ068114.1 | - | 3 | 555 | |||
CD048295.1 | - | 3 | 773 | |||
M38188.1 | - | 1 | 726 |