Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153240.4 | NP_694972.3 | 7 | 1286 | ||
GenBank mRNA | AK055253.1 | - | 2 | 229 | ||
AL832877.1 | - | 6 | 3317 | |||
AY257864.1 | - | 7 | 1215 | |||
AY257866.1 | - | 8 | 997 | |||
BC131729.1 | - | 7 | 937 | |||
CR749498.1 | - | 4 | 958 |
Gene Symbol: NPHP3-ACAD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037804.1 | 7 | 1286 | |||
GenBank mRNA | AB082531.1 | - | 7 | 1286 |