Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014383.2 | NP_055198.1 | 3 | 847 | ||
XM_011526718.2 | XP_011525020.1 | 2 | 1008 | |||
XM_017026588.1 | XP_016882077.1 | 1 | 513 | |||
XM_017026589.1 | XP_016882078.1 | 3 | 1218 | |||
XM_017026590.1 | XP_016882079.1 | 1 | 513 | |||
GenBank mRNA | AF130255.1 | - | 2 | 699 | ||
AF165097.1 | - | 1 | 489 | |||
BC017700.2 | - | 2 | 615 | 4419776 | ||
KU178599.1 | - | 1 | 489 | |||
KU178600.1 | - | 1 | 489 |