Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316902.1 | NP_001303831.1 | 7 | 623 | ||
NM_001316903.1 | NP_001303832.1 | 6 | 549 | |||
NM_014383.2 | NP_055198.1 | 7 | 1563 | |||
XM_011526718.2 | XP_011525020.1 | 6 | 1724 | |||
XM_017026588.1 | XP_016882077.1 | 4 | 1337 | |||
XM_017026589.1 | XP_016882078.1 | 7 | 1934 | |||
XM_017026590.1 | XP_016882079.1 | 3 | 1335 | |||
XM_017026591.1 | XP_016882080.1 | 6 | 737 | |||
XM_017026592.1 | XP_016882081.1 | 4 | 417 | |||
GenBank mRNA | AF130255.1 | - | 6 | 1415 | ||
AF165097.1 | - | 5 | 1205 | |||
BC017700.2 | - | 4 | 1188 | 4419776 |