Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014390.2 | NP_055205.2 | 5 | 756 | ||
XM_017011987.1 | XP_016867476.1 | 5 | 785 | |||
GenBank mRNA | AB209510.1 | - | 5 | 694 | ||
AK096583.1 | - | 6 | 681 | |||
AK304181.1 | - | 5 | 648 | |||
AK312313.1 | - | 5 | 710 | |||
BC017180.2 | - | 5 | 712 | 3345037 | ||
BT009785.1 | - | 5 | 530 | |||
U22055.1 | - | 5 | 722 |