Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014391.2 | NP_055206.2 | 5, 6 | 793 | ||
GenBank mRNA | AK290960.1 | - | 5, 6 | 605 | ||
AY903446.1 | - | 5, 6 | 562 | |||
BC018667.1 | - | 5, 6 | 609 | 4245591 | ||
X83703.1 | - | 5, 6 | 794 |