Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306151.1 | NP_001293080.1 | 7 | 730 | ||
NM_014395.2 | NP_055210.2 | 7 | 739 | |||
XM_011531840.2 | XP_011530142.1 | 7 | 733 | |||
XM_011531842.1 | XP_011530144.1 | 7 | 733 | |||
XM_011531843.2 | XP_011530145.1 | 5 | 528 | |||
XM_017008023.1 | XP_016863512.1 | 7 | 733 | |||
XM_017008024.1 | XP_016863513.1 | 7 | 733 | |||
GenBank mRNA | AF161551.1 | - | 7 | 714 | ||
AF163254.1 | - | 7 | 720 | |||
AF178987.1 | - | 7 | 694 | |||
AF186022.1 | - | 7 | 730 | |||
AK301355.1 | - | 6 | 582 | |||
BC012924.1 | - | 7 | 664 | 4517170 |