Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014400.2 | NP_055215.2 | 2 | 253 | ||
GenBank mRNA | AF082889.1 | - | 2 | 230 | ||
AJ223603.2 | - | 2 | 181 | |||
AK075464.1 | - | 2 | 182 | |||
AK290905.1 | - | 2 | 182 | |||
AK314773.1 | - | 2 | 182 | |||
AL832153.1 | - | 2 | 183 | |||
AY359006.1 | - | 2 | 160 | |||
BC039167.1 | - | 2 | 167 | 4751561 | ||
BC041061.1 | - | 2 | 162 | |||
BP367221.1 | - | 2 | 227 | |||
DA837818.1 | - | 2 | 253 |