Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022045.4 | NP_071328.2 | 18 | 2272 | ||
GenBank mRNA | BC013136.1 | - | 18 | 2244 | 4476886 |