Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018644.3 | NP_061114.2 | 7 | 1348 | ||
NM_054025.2 | NP_473366.1 | 6 | 1480 | |||
XM_005271506.3 | XP_005271563.1 | 7 | 1399 | |||
XM_011542751.2 | XP_011541053.1 | 8 | 1611 | |||
XM_017017551.1 | XP_016873040.1 | 7 | 3173 | |||
XM_017017552.1 | XP_016873041.1 | 5 | 1935 | |||
GenBank mRNA | AB029396.1 | - | 7 | 1262 | ||
AL834455.1 | - | 4 | 656 | |||
BC010466.1 | - | 6 | 1480 | 4157589 |