Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCRQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014402.4 | NP_055217.2 | 3 | 278 | ||
GenBank mRNA | AK297061.1 | - | 2 | 715 | ||
AK312148.1 | - | 3 | 248 | |||
AW327741.1 | - | 2 | 169 | |||
BC001390.2 | - | 2 | 231 | 3050489 | ||
BC064949.1 | - | 3 | 252 | |||
BC090048.1 | - | 3 | 255 | 30393278 | ||
BU953465.1 | - | 3 | 281 |