Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006612.5 | NP_006603.2 | 10, 11 | 1221 | ||
XM_005256424.2 | XP_005256481.1 | 11, 12 | 1212 | |||
GenBank mRNA | AB014606.2 | - | 10, 11 | 1217 | ||
AK307401.1 | - | 10, 11 | 1064 | |||
BC034993.1 | - | 10, 11 | 1195 | 5266798 | ||
U91329.1 | - | 9, 10 | 977 |