Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014415.3 | NP_055230.2 | 11 | 3248 | ||
XM_011512689.2 | XP_011510991.1 | 11 | 2956 | |||
XM_011512690.2 | XP_011510992.1 | 9 | 4633 | |||
GenBank mRNA | AB208862.1 | - | 8 | 1961 | ||
AK023488.1 | - | 3 | 700 | |||
AK312859.1 | - | 11 | 3240 | |||
BC111700.1 | - | 11 | 3155 | 40038346 | ||
U69274.1 | - | 11 | 3179 |