Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM155B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015686.2 | NP_056501.2 | 3 | 1208 | ||
GenBank mRNA | AF087142.1 | - | 3 | 1210 | ||
BC136518.1 | - | 3 | 1195 | 9020508 | ||
BC136519.1 | - | 3 | 1195 | 9020509 |