Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318381.1 | NP_001305310.1 | 11 | 1742 | ||
NM_001318382.1 | NP_001305311.1 | 10 | 1665 | |||
NM_014425.4 | NP_055240.2 | 10 | 1654 | |||
NR_134606.1 | 10 | 1654 | ||||
GenBank mRNA | AF039217.2 | - | 10 | 1605 | ||
AF084367.1 | - | 9 | 1424 | |||
AK304851.1 | - | 11 | 1664 | |||
AK312795.1 | - | 10 | 1607 | |||
BC041665.1 | - | 10 | 1610 | |||
BC056897.1 | - | 12 | 1777 | |||
BC111761.1 | - | 9 | 1437 | 40012263 | ||
BC112351.1 | - | 9 | 1437 | 40012265 |