Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145873.1 | NP_001139345.1 | 9 | 2460 | ||
NM_001768.6 | NP_001759.3 | 6 | 2318 | |||
NM_171827.3 | NP_741969.1 | 5 | 2207 | |||
NR_027353.1 | 7 | 1904 | ||||
GenBank mRNA | AK124156.1 | - | 9 | 2460 | ||
BC025715.1 | - | 6 | 1433 | 5227906 | ||
M12824.1 | - | 6 | 1516 |