Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171999.3 | NP_741996.2 | 3 | 3871 | ||
GenBank mRNA | BC148296.2 | - | 5 | 2848 | 40114022 |