Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280796.1 | NP_001267725.1 | 12 | 1436 | ||
NM_001280797.1 | NP_001267726.1 | 11 | 1233 | |||
NM_001280798.1 | NP_001267727.1 | 9 | 1011 | |||
NM_013267.3 | NP_037399.2 | 12 | 1363 | |||
XM_005268797.1 | XP_005268854.1 | 12 | 990 | |||
XM_017019180.1 | XP_016874669.1 | 11 | 860 | |||
GenBank mRNA | AF110330.1 | - | 12 | 1118 | ||
AF110331.1 | - | 2 | 50 | |||
AF223944.1 | - | 12 | 1290 | |||
AK000467.1 | - | 12 | 1110 | |||
AK303772.1 | - | 11 | 1499 | |||
BC025661.1 | - | 11 | 948 | |||
HM775422.1 | - | 11 | 982 | |||
HM775423.1 | - | 10 | 852 | |||
HM775424.1 | - | 10 | 915 |