Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013245.2 | NP_037377.1 | 3 | 340 | ||
GenBank mRNA | AF112215.1 | - | 3 | 295 | ||
AF132747.1 | - | 3 | 348 | |||
AF155740.1 | - | 2 | 198 | |||
AF159063.1 | - | 3 | 340 | |||
AF255952.1 | - | 3 | 340 | |||
AK315026.1 | - | 3 | 368 | |||
BC047932.1 | - | 3 | 318 | 5753632 |