Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSNAX-DISC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028393.1 | 6 | 1342 | |||
NR_028394.1 | 7 | 1470 | ||||
NR_028395.1 | 7 | 1470 | ||||
NR_028396.1 | 6 | 1342 | ||||
NR_028397.1 | 6 | 1207 | ||||
NR_028398.1 | 4 | 961 | ||||
NR_028399.1 | 8 | 1577 | ||||
NR_028400.1 | 7 | 1449 |