Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001769.3 | NP_001760.1 | 7 | 750 | ||
XM_011521040.1 | XP_011519342.1 | 7 | 729 | |||
GenBank mRNA | AB079244.1 | - | 7 | 671 | ||
AK298431.1 | - | 6 | 380 | |||
AK315219.1 | - | 7 | 699 | |||
AY423720.1 | - | 7 | 583 | |||
AY966455.1 | - | 7 | 678 | |||
BC011988.1 | - | 7 | 617 | 3860667 | ||
L34068.1 | - | 7 | 566 | |||
M38690.1 | - | 7 | 617 | |||
X60111.1 | - | 7 | 677 |