Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001769.3 | NP_001760.1 | 2 | 323 | ||
XM_005253814.3 | XP_005253871.1 | 2 | 320 | |||
XM_011521040.1 | XP_011519342.1 | 2 | 302 | |||
GenBank mRNA | AB079244.1 | - | 2 | 244 | ||
AK296332.1 | - | 2 | 266 | |||
AK296894.1 | - | 2 | 239 | |||
AK308700.1 | - | 2 | 270 | |||
AK311020.1 | - | 2 | 302 | |||
AK315219.1 | - | 2 | 272 | |||
AY423720.1 | - | 2 | 156 | |||
AY966455.1 | - | 2 | 250 | |||
BC011988.1 | - | 2 | 190 | 3860667 | ||
BG291377.1 | - | 2 | 325 | |||
BM763066.1 | - | 2 | 323 | |||
L34068.1 | - | 2 | 139 | |||
M38690.1 | - | 2 | 190 | |||
X60111.1 | - | 2 | 250 |