Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001769.3 | NP_001760.1 | 6 | 673 | ||
XM_005253814.3 | XP_005253871.1 | 5 | 818 | |||
XM_011521040.1 | XP_011519342.1 | 6 | 652 | |||
GenBank mRNA | AB079244.1 | - | 6 | 594 | ||
AK296332.1 | - | 5 | 764 | |||
AK298431.1 | - | 5 | 303 | |||
AK308700.1 | - | 5 | 666 | |||
AK309527.1 | - | 2 | 418 | |||
AK315219.1 | - | 6 | 622 | |||
AY423720.1 | - | 6 | 506 | |||
AY966455.1 | - | 6 | 601 | |||
BC011988.1 | - | 6 | 540 | 3860667 | ||
L34068.1 | - | 6 | 489 | |||
M38690.1 | - | 6 | 540 | |||
X60111.1 | - | 6 | 600 |