Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286414.2 | NP_001273343.1 | 15 | 1933 | ||
NM_014444.4 | NP_055259.2 | 15 | 1930 | |||
XM_011521454.2 | XP_011519756.1 | 15 | 1933 | |||
XM_011521455.2 | XP_011519757.1 | 15 | 1831 | |||
XM_017022078.1 | XP_016877567.1 | 15 | 1930 | |||
GenBank mRNA | AJ249677.1 | - | 15 | 1639 | ||
AK001639.1 | - | 11 | 1244 | |||
AK026551.1 | - | 15 | 1920 | |||
AK027703.1 | - | 15 | 1891 | |||
AK127306.1 | - | 5 | 2074 | |||
BC000966.2 | - | 14 | 1456 | 3446803 | ||
BC009870.2 | - | 15 | 1855 | 3940649 | ||
BC012801.2 | - | 15 | 1874 | 2821891 |