Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318865.1 | NP_001305794.1 | 4 | 477 | ||
NM_018960.5 | NP_061833.1 | 4 | 534 | |||
GenBank mRNA | AF101477.1 | - | 4 | 530 | ||
BC032627.1 | - | 4 | 526 | 5229272 | ||
FJ224320.1 | - | 4 | 530 | |||
KU178603.1 | - | 4 | 520 | |||
KU178604.1 | - | 4 | 463 |
Gene Symbol: CNPY3-GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318856.1 | NP_001305785.1 | 4 | 906 | ||
NM_001318857.1 | NP_001305786.1 | 3 | 778 | |||
GenBank mRNA | BQ947041.1 | - | 4 | 533 | ||
FY210916.1 | - | 3 | 311 |