Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPY3-GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318856.1 | NP_001305785.1 | 4, 5 | 973 | ||
NM_001318857.1 | NP_001305786.1 | 3, 4 | 845 | |||
NM_001318858.1 | NP_001305787.1 | 3, 4 | 737 | |||
GenBank mRNA | BQ947041.1 | - | 4, 5 | 601 | ||
FY210916.1 | - | 3, 4 | 378 | |||
FY213777.1 | - | 3, 4 | 270 |
Gene Symbol: GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318865.1 | NP_001305794.1 | 4, 5 | 544 | ||
NM_018960.5 | NP_061833.1 | 4, 5 | 601 | |||
GenBank mRNA | AF101477.1 | - | 4, 5 | 597 | ||
BC032627.1 | - | 4, 5 | 593 | 5229272 | ||
FJ224320.1 | - | 4, 5 | 597 | |||
KU178603.1 | - | 4, 5 | 587 | |||
KU178604.1 | - | 4, 5 | 530 | |||
X62250.1 | - | 4, 5 | 735 |