Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318865.1 | NP_001305794.1 | 5 | 585 | ||
NM_018960.5 | NP_061833.1 | 5 | 642 | |||
NR_134899.1 | 4 | 499 | ||||
GenBank mRNA | AF101477.1 | - | 5 | 638 | ||
BC032627.1 | - | 5 | 634 | 5229272 | ||
FJ224320.1 | - | 5 | 638 | |||
KU178603.1 | - | 5 | 628 | |||
KU178604.1 | - | 5 | 571 | |||
KU178605.1 | - | 4 | 485 | |||
X62250.1 | - | 5 | 776 |
Gene Symbol: CNPY3-GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318856.1 | NP_001305785.1 | 5 | 1014 | ||
NM_001318857.1 | NP_001305786.1 | 4 | 886 | |||
NM_001318858.1 | NP_001305787.1 | 4 | 778 | |||
NR_134890.1 | 4 | 840 | ||||
NR_134891.1 | 3 | 743 | ||||
NR_134892.1 | 4 | 871 | ||||
GenBank mRNA | FY210916.1 | - | 4 | 419 | ||
FY213370.1 | - | 3 | 275 | |||
FY213777.1 | - | 4 | 311 | |||
FY213816.1 | - | 4 | 372 |